Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161009.2 | NP_001154481.1 | 3 | 711 | ||
NM_001161010.2 | NP_001154482.1 | 4 | 816 | |||
NM_001161011.2 | NP_001154483.1 | 3 | 711 | |||
NM_001161012.2 | NP_001154484.1 | 3 | 711 | |||
NM_001161013.2 | NP_001154485.1 | 3 | 711 | |||
NM_031905.4 | NP_114111.2 | 4 | 816 | |||
XM_005250628.2 | XP_005250685.1 | 4 | 970 | |||
XM_011516601.2 | XP_011514903.1 | 3 | 889 | |||
XM_011516602.2 | XP_011514904.1 | 3 | 250 | |||
XM_017012684.1 | XP_016868173.1 | 5 | 997 | |||
XM_017012685.1 | XP_016868174.1 | 5 | 997 | |||
XM_017012686.1 | XP_016868175.1 | 5 | 997 | |||
XM_017012687.1 | XP_016868176.1 | 3 | 229 | |||
XM_017012688.1 | XP_016868177.1 | 5 | 997 | |||
GenBank mRNA | AF275808.1 | - | 3 | 219 | ||
AK075500.1 | - | 4 | 557 | |||
AK225082.1 | - | 3 | 444 | |||
AK291818.1 | - | 3 | 439 | |||
AK301565.1 | - | 3 | 444 | |||
AY150851.1 | - | 3 | 711 | |||
AY150852.1 | - | 4 | 816 | |||
AY150853.1 | - | 3 | 711 | |||
AY150854.1 | - | 4 | 816 | |||
BC003586.1 | - | 3 | 431 | 3357276 |