Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETDB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160308.2 | NP_001153780.1 | 6 | 1707 | ||
NM_001320699.1 | NP_001307628.1 | 6 | 1386 | |||
NM_031915.2 | NP_114121.2 | 7 | 1743 | |||
GenBank mRNA | AB209349.1 | - | 1 | 159 | ||
AF334407.1 | - | 7 | 1662 | |||
AK307593.1 | - | 5 | 1056 | |||
AK308220.1 | - | 6 | 1626 | |||
AK308354.1 | - | 5 | 1203 | |||
BC028202.1 | - | 5 | 1180 | |||
BC047434.2 | - | 5 | 1143 | 5266911 |
Gene Symbol: SETDB2-PHF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320727.1 | NP_001307656.1 | 6 | 1707 | ||
NR_135324.1 | 6 | 1707 |