Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031923.3 | NP_114129.1 | 3 | 1585 | ||
XM_011519741.1 | XP_011518043.1 | 3 | 1609 | |||
GenBank mRNA | AJ292190.1 | - | 3 | 1379 | ||
AK307521.1 | - | 3 | 1558 | |||
AK307657.1 | - | 3 | 1563 | |||
AL117661.1 | - | 3 | 1313 | |||
BC017320.1 | - | 3 | 1571 | |||
BC028077.1 | - | 3 | 1542 | |||
BC045106.1 | - | 3 | 1571 | |||
BC062352.1 | - | 3 | 1552 | |||
BC073884.1 | - | 3 | 1576 |