Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127174.1 | NP_001120646.1 | 5 | 539 | ||
NM_001127175.1 | NP_001120647.1 | 5 | 650 | |||
NM_001127176.1 | NP_001120648.1 | 6 | 806 | |||
NM_031939.3 | NP_114145.2 | 7 | 909 | |||
XM_005258365.3 | XP_005258422.1 | 6 | 719 | |||
XM_011526226.2 | XP_011524528.1 | 6 | 690 | |||
XM_017026035.1 | XP_016881524.1 | 6 | 686 | |||
XM_017026036.1 | XP_016881525.1 | 6 | 690 | |||
XM_017026037.1 | XP_016881526.1 | 5 | 679 | |||
GenBank mRNA | AB042647.1 | - | 7 | 773 | ||
AK054702.1 | - | 7 | 843 | |||
AK294757.1 | - | 5 | 494 | |||
AK295666.1 | - | 5 | 651 | |||
BC029860.1 | - | 7 | 796 | 5171860 | ||
DB473056.1 | - | 4 | 388 | |||
DC403805.1 | - | 5 | 535 |