Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDCA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031942.4 | NP_114148.3 | 7 | 1151 | ||
NM_145810.2 | NP_665809.1 | 6 | 914 | |||
XM_011511957.2 | XP_011510259.1 | 7 | 1692 | |||
GenBank mRNA | AK027628.1 | - | 7 | 1123 | ||
AK027642.1 | - | 6 | 895 | |||
AK075134.1 | - | 5 | 1777 | |||
AK223524.1 | - | 6 | 919 | |||
AK297097.1 | - | 5 | 749 | |||
AK297245.1 | - | 7 | 1011 | |||
AK300949.1 | - | 7 | 980 | |||
AL833728.1 | - | 2 | 443 | |||
AY029179.1 | - | 6 | 927 | |||
BC015124.1 | - | 2 | 330 | |||
BC027966.1 | - | 6 | 880 | 5227112 |