Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290212.1 | NP_001277141.1 | 3 | 782 | ||
NM_031945.4 | NP_114151.3 | 2 | 548 | |||
NR_110866.1 | 2 | 548 | ||||
GenBank mRNA | BC021923.1 | - | 2 | 517 | 4876887 | |
BG760374.1 | - | 2 | 517 |