Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031952.3 | NP_114158.2 | 2 | 291 | ||
NR_125327.1 | 2 | 291 | ||||
NR_125328.1 | 2 | 266 | ||||
NR_125329.1 | 2 | 291 | ||||
NR_125330.1 | 2 | 291 | ||||
XM_011543664.1 | XP_011541966.1 | 2 | 247 | |||
XM_011543666.1 | XP_011541968.1 | 2 | 291 | |||
XM_017009948.1 | XP_016865437.1 | 2 | 272 | |||
XM_017009949.1 | XP_016865438.1 | 7 | 828 | |||
XM_017009950.1 | XP_016865439.1 | 6 | 810 | |||
GenBank mRNA | AK093225.1 | - | 2 | 291 | ||
AK292338.1 | - | 2 | 272 | |||
AK310260.1 | - | 2 | 247 | |||
AY027526.1 | - | 2 | 240 | |||
BC032832.1 | - | 2 | 296 | 4827641 | ||
BC047333.1 | - | 2 | 277 | 5166271 | ||
DB027274.1 | - | 2 | 291 |