Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282150.1 | NP_001269079.1 | 4 | 378 | ||
NM_001282151.1 | NP_001269080.1 | 2 | 223 | |||
NM_016226.4 | NP_057310.1 | 2 | 223 | |||
NM_057180.2 | NP_476528.1 | 3 | 235 | |||
NR_104099.1 | 4 | 275 | ||||
NR_104100.1 | 3 | 263 | ||||
NR_104101.1 | 3 | 313 | ||||
NR_104102.1 | 2 | 114 | ||||
XM_006719459.3 | XP_006719522.1 | 4 | 284 | |||
XM_006719460.3 | XP_006719523.1 | 5 | 321 | |||
XM_011538484.2 | XP_011536786.1 | 3 | 269 | |||
XM_017019457.1 | XP_016874946.1 | 3 | 294 | |||
XM_017019458.1 | XP_016874947.1 | 2 | 414 | |||
XM_017019459.1 | XP_016874948.1 | 3 | 312 | |||
GenBank mRNA | AA148059.1 | - | 2 | 61 | ||
AB578891.1 | - | 3 | 204 | |||
AB578892.1 | - | 3 | 235 | |||
AB618733.1 | - | 2 | 206 | |||
AF086379.1 | - | 1 | 129 | |||
AF168716.1 | - | 3 | 193 | |||
AF175264.1 | - | 2 | 192 | |||
AF193795.1 | - | 2 | 185 | |||
AF201936.1 | - | 3 | 280 | |||
AF201946.1 | - | 2 | 171 | |||
AK129767.1 | - | 2 | 185 | |||
AK130510.1 | - | 1021 | ||||
AL832866.1 | - | 2 | 2216 | |||
BC000880.1 | - | 3 | 193 | 3461977 | ||
BC015095.1 | - | 3 | 202 | |||
BC095446.1 | - | 2 | 194 | 30719438 | ||
BF030144.1 | - | 234 | ||||
BG707325.1 | - | 2 | 218 | |||
BM806646.1 | - | 4 | 331 | |||
BU568689.1 | - | 4 | 236 | |||
CB158821.1 | - | 2 | 157 | |||
CR457182.1 | - | 1 | 146 |