Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD3NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032016.3 | NP_114405.1 | 5 | 693 | ||
XM_005249880.1 | XP_005249937.1 | 6 | 707 | |||
XM_011515572.1 | XP_011513874.1 | 5 | 628 | |||
XM_017012692.1 | XP_016868181.1 | 5 | 726 | |||
XM_017012693.1 | XP_016868182.1 | 6 | 657 | |||
XM_017012694.1 | XP_016868183.1 | 5 | 688 | |||
GenBank mRNA | AJ492267.1 | - | 5 | 568 | ||
AK091579.1 | - | 4 | 2498 | |||
AK315404.1 | - | 5 | 662 | |||
AY358645.1 | - | 5 | 519 | |||
BC003074.2 | - | 4 | 478 | 3505985 | ||
BC005959.1 | - | 5 | 513 | 4048871 |