Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282546.1 | NP_001269475.1 | 8 | 1097 | ||
NM_001282547.1 | NP_001269476.1 | 8 | 1082 | |||
NM_032017.2 | NP_114406.1 | 9 | 1319 | |||
GenBank mRNA | AK024504.1 | - | 7 | 881 | ||
AK075029.1 | - | 5 | 598 | |||
AK075048.1 | - | 8 | 1076 | |||
AK131560.1 | - | 1 | 1459 | |||
AL834137.1 | - | 9 | 1276 | |||
AY198395.1 | - | 8 | 1052 | |||
BC005169.2 | - | 3 | 281 | |||
BC007835.2 | - | 8 | 1050 | 4109652 | ||
BC008344.1 | - | 8 | 1058 | 3530483 |