Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032027.2 | NP_114416.1 | 6 | 886 | ||
NR_135160.1 | 7 | 1016 | ||||
NR_135161.1 | 5 | 812 | ||||
NR_135162.1 | 7 | 1013 | ||||
GenBank mRNA | AF353990.1 | - | 6 | 886 | ||
AK311444.1 | - | 6 | 532 | |||
AK311705.1 | - | 7 | 663 | |||
BC029486.1 | - | 6 | 604 | 5271098 | ||
BC048995.1 | - | 6 | 602 |