Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCALD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040624.1 | NP_001035714.1 | 7, 8 | 870 | ||
NM_001040625.1 | NP_001035715.1 | 6, 7 | 783 | |||
NM_001040626.1 | NP_001035716.1 | 6, 7 | 783 | |||
NM_001040627.1 | NP_001035717.1 | 5, 6 | 696 | |||
NM_001040628.1 | NP_001035718.1 | 7, 8 | 839 | |||
NM_001040629.1 | NP_001035719.1 | 6, 7 | 752 | |||
NM_001040630.1 | NP_001035720.1 | 5, 6 | 936 | |||
NM_032041.2 | NP_114430.2 | 3, 4 | 845 | |||
XM_011517332.2 | XP_011515634.1 | 3, 4 | 634 | |||
XM_011517333.2 | XP_011515635.1 | 7, 8 | 781 | |||
XM_011517334.2 | XP_011515636.1 | 4, 5 | 665 | |||
XM_011517335.2 | XP_011515637.1 | 5, 6 | 757 | |||
XM_017013901.1 | XP_016869390.1 | 6, 7 | 973 | |||
XM_017013902.1 | XP_016869391.1 | 4, 5 | 644 | |||
GenBank mRNA | AF251061.1 | - | 5, 6 | 624 | ||
AK055073.1 | - | 3, 4 | 603 | |||
AK055097.1 | - | 1, 2 | 1573 | |||
AK096515.1 | - | 7, 8 | 795 | |||
AK314523.1 | - | 3, 4 | 604 | |||
AL136612.1 | - | 3, 4 | 596 | |||
BC036098.1 | - | 5, 6 | 623 | 5274150 | ||
BC063428.1 | - | 6, 7 | 769 | 5240958 |