Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM172A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032042.5 | NP_114431.2 | 2, 3 | 307 | ||
NR_028080.1 | 2, 3 | 307 | ||||
XM_005272103.3 | XP_005272160.1 | 2, 3 | 273 | |||
XM_005272105.3 | XP_005272162.1 | 2, 3 | 272 | |||
XM_005272106.3 | XP_005272163.1 | 2, 3 | 273 | |||
XM_005272108.3 | XP_005272165.1 | 2, 3 | 302 | |||
XM_005272109.4 | XP_005272166.1 | 2, 3 | 273 | |||
XM_005272111.4 | XP_005272168.1 | 2, 3 | 273 | |||
XM_006714717.3 | XP_006714780.1 | 2, 3 | 299 | |||
XM_011543668.2 | XP_011541970.1 | 2, 3 | 273 | |||
XM_011543670.2 | XP_011541972.1 | 2, 3 | 273 | |||
XM_011543671.2 | XP_011541973.1 | 2, 3 | 273 | |||
XM_011543674.2 | XP_011541976.1 | 2, 3 | 273 | |||
XM_017009954.1 | XP_016865443.1 | 2, 3 | 273 | |||
XM_017009961.1 | XP_016865450.1 | 2, 3 | 217 | |||
GenBank mRNA | AK295127.1 | - | 2, 3 | 307 | ||
AK297471.1 | - | 1, 2 | 69 | |||
AK312930.1 | - | 2, 3 | 212 | |||
AL136630.1 | - | 2, 3 | 146 | |||
BC020584.2 | - | 1, 2 | 109 | 4398813 | ||
DC324758.1 | - | 2, 3 | 307 | |||
DC374283.1 | - | 2, 3 | 292 |