Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM172A SIRNA Assays ›
Gene Symbol:
FAM172A
Gene Name:
family with sequence similarity 172 member A
Gene Aliases:
C5orf21
Chromosome Location:
Chr.5: 93615130 - 94111699 on Build GRCh38
UniGene ID:
Hs.600086
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s38382
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM172A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001163417.1
NP_001156889.1
8
1048
NM_001163418.1
NP_001156890.1
7
856
NM_032042.5
NP_114431.2
9
1172
NR_028080.1
7
879
XM_005272103.3
XP_005272160.1
9
1138
XM_005272105.3
XP_005272162.1
8
945
XM_005272106.3
XP_005272163.1
8
919
XM_005272108.3
XP_005272165.1
8
1066
XM_005272110.3
XP_005272167.1
7
773
XM_006714717.3
XP_006714780.1
9
1164
XM_006714718.3
XP_006714781.1
7
1778
XM_006714719.3
XP_006714782.1
7
927
XM_011543668.2
XP_011541970.1
9
1138
XM_011543670.2
XP_011541972.1
9
1138
XM_011543671.2
XP_011541973.1
9
1138
XM_011543672.2
XP_011541974.1
9
1068
XM_011543673.2
XP_011541975.1
8
967
XM_017009952.1
XP_016865441.1
9
1460
XM_017009953.1
XP_016865442.1
9
1458
XM_017009954.1
XP_016865443.1
9
1138
XM_017009955.1
XP_016865444.1
8
1876
XM_017009956.1
XP_016865445.1
8
1190
XM_017009957.1
XP_016865446.1
7
1144
XM_017009958.1
XP_016865447.1
7
939
XM_017009960.1
XP_016865449.1
6
721
GenBank mRNA
AK295127.1
-
8
1071
AK295879.1
-
7
805
AK296683.1
-
4
442
AK296986.1
-
8
1025
AK297471.1
-
6
641
AK312930.1
-
9
1077
BC020584.2
-
8
974
4398813
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001106401.
This siRNA also targets the following transcripts in Mouse: NM_001163420, NM_001163419, NM_138312, AF064782, BC057380, and others.
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-8tsm6:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline