Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282150.1 | NP_001269079.1 | 5 | 613 | ||
NM_016226.4 | NP_057310.1 | 3 | 458 | |||
NM_057180.2 | NP_476528.1 | 4 | 470 | |||
NR_104099.1 | 5 | 510 | ||||
NR_104100.1 | 4 | 498 | ||||
NR_104101.1 | 4 | 548 | ||||
NR_104102.1 | 3 | 349 | ||||
XM_006719459.3 | XP_006719522.1 | 5 | 519 | |||
XM_006719460.3 | XP_006719523.1 | 6 | 556 | |||
XM_011538484.2 | XP_011536786.1 | 4 | 504 | |||
XM_017019457.1 | XP_016874946.1 | 4 | 529 | |||
XM_017019458.1 | XP_016874947.1 | 3 | 649 | |||
GenBank mRNA | AA148059.1 | - | 3 | 296 | ||
AB578890.1 | - | 2 | 197 | |||
AF168716.1 | - | 4 | 428 | |||
AF175264.1 | - | 3 | 427 | |||
AF193795.1 | - | 3 | 420 | |||
AF201936.1 | - | 4 | 515 | |||
AF201946.1 | - | 3 | 406 | |||
AK000499.1 | - | 2 | 186 | |||
AK129767.1 | - | 3 | 420 | |||
AK130510.1 | - | 1256 | ||||
AL136614.1 | - | 3 | 436 | |||
AL832866.1 | - | 3 | 2451 | |||
BC000880.1 | - | 4 | 428 | 3461977 | ||
BC015095.1 | - | 4 | 437 | |||
BC095446.1 | - | 3 | 429 | 30719438 | ||
BF030144.1 | - | 468 | ||||
BG707325.1 | - | 3 | 453 | |||
BM806646.1 | - | 5 | 566 | |||
BU568689.1 | - | 5 | 471 | |||
CR457182.1 | - | 2 | 381 | |||
CR533468.1 | - | 2 | 369 |