Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320823.1 | NP_001307752.1 | 2 | 162 | ||
NM_001320824.1 | NP_001307753.1 | 2 | 217 | |||
NM_032118.3 | NP_115494.1 | 2 | 184 | |||
GenBank mRNA | AK023015.1 | - | 2 | 177 | ||
AK222696.1 | - | 2 | 177 | |||
BC051848.1 | - | 2 | 149 | 6150670 | ||
BG024606.1 | - | 2 | 183 | |||
BI855898.1 | - | 2 | 125 | |||
HY088750.1 | - | 2 | 199 |