Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032119.3 | NP_115495.3 | 13 | 2575 | ||
NR_003149.1 | 13 | 2575 | ||||
XM_017009963.1 | XP_016865452.1 | 13 | 2694 | |||
XM_017009964.1 | XP_016865453.1 | 13 | 2694 | |||
XM_017009965.1 | XP_016865454.1 | 13 | 2576 | |||
XM_017009966.1 | XP_016865455.1 | 13 | 2694 | |||
XM_017009967.1 | XP_016865456.1 | 12 | 2483 | |||
XM_017009968.1 | XP_016865457.1 | 13 | 2694 | |||
XM_017009969.1 | XP_016865458.1 | 13 | 2694 | |||
XM_017009970.1 | XP_016865459.1 | 13 | 2694 | |||
XM_017009971.1 | XP_016865460.1 | 13 | 2694 | |||
XM_017009974.1 | XP_016865463.1 | 13 | 2694 | |||
GenBank mRNA | AB075823.1 | - | 6 | 1245 | ||
AF435925.1 | - | 13 | 2575 |