Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIRREL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032123.6 | NP_115499.5 | 5 | 634 | ||
NM_199179.3 | NP_954648.3 | 4 | 484 | |||
NM_199180.3 | NP_954649.3 | 5 | 776 | |||
XM_011527362.1 | XP_011525664.1 | 6 | 836 | |||
XM_011527363.1 | XP_011525665.1 | 6 | 817 | |||
XM_011527365.1 | XP_011525667.1 | 5 | 686 | |||
XM_011527366.1 | XP_011525668.1 | 4 | 947 | |||
XM_017027348.1 | XP_016882837.1 | 5 | 1343 | |||
GenBank mRNA | AB593116.1 | - | 5 | 740 | ||
AK307865.1 | - | 5 | 1097 | |||
AL136654.1 | - | 5 | 759 | |||
AY305301.1 | - | 5 | 578 | |||
AY305302.1 | - | 4 | 428 | |||
AY358742.1 | - | 5 | 762 |