Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318765.1 | NP_001305694.1 | 3 | 474 | ||
NM_032124.4 | NP_115500.1 | 3 | 417 | |||
XM_005258371.4 | XP_005258428.1 | 3 | 437 | |||
XM_006722557.2 | XP_006722620.1 | 4 | 509 | |||
XM_011526227.2 | XP_011524529.1 | 4 | 493 | |||
XM_011526228.2 | XP_011524530.1 | 4 | 491 | |||
XM_017026039.1 | XP_016881528.1 | 4 | 530 | |||
XM_017026040.1 | XP_016881529.1 | 4 | 509 | |||
GenBank mRNA | AK054590.1 | - | 3 | 454 | ||
AK292098.1 | - | 3 | 417 | |||
AL136681.1 | - | 3 | 408 | |||
BC011894.2 | - | 3 | 438 | 3944181 | ||
BC033031.1 | - | 3 | 453 | 4793566 | ||
BC038100.2 | - | 3 | 417 | 5266439 | ||
CR533487.1 | - | 2 | 284 | |||
FJ224322.1 | - | 3 | 408 |