Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032125.2 | NP_115501.2 | 2 | 297 | ||
NR_037576.1 | 2 | 297 | ||||
NR_037577.1 | 2 | 297 | ||||
NR_037580.1 | 2 | 297 | ||||
GenBank mRNA | AF251699.1 | - | 2 | 193 | ||
AK222642.1 | - | 2 | 176 | |||
AK303821.1 | - | 2 | 251 | |||
AK307077.1 | - | 2 | 135 | |||
AL136683.1 | - | 2 | 211 | |||
BC011579.1 | - | 2 | 194 | |||
BC090039.1 | - | 2 | 189 | 6181849 | ||
BP326836.1 | - | 2 | 282 | |||
BQ927627.1 | - | 2 | 102 | |||
BX394857.2 | - | 2 | 157 | |||
DC424669.1 | - | 2 | 299 |