Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170722.1 | NP_001164193.1 | 5 | 398 | ||
NM_001170723.1 | NP_001164194.1 | 5 | 374 | |||
NM_001170724.1 | NP_001164195.1 | 5 | 374 | |||
NM_032126.4 | NP_115502.2 | 5 | 374 | |||
GenBank mRNA | AK093269.1 | - | 5 | 321 | ||
AL136694.1 | - | 5 | 378 | |||
AL552172.3 | - | 5 | 297 | |||
BC034972.2 | - | 5 | 331 | 4823687 | ||
BI461686.1 | - | 5 | 338 | |||
BI825610.1 | - | 5 | 308 |