Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HORMAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199829.1 | NP_001186758.1 | 3 | 236 | ||
NM_032132.4 | NP_115508.2 | 3 | 236 | |||
XM_006711570.2 | XP_006711633.1 | 3 | 194 | |||
XM_006711571.2 | XP_006711634.1 | 1 | 77 | |||
XM_011510053.2 | XP_011508355.1 | 3 | 304 | |||
XM_011510054.2 | XP_011508356.1 | 3 | 258 | |||
GenBank mRNA | AK097390.1 | - | 3 | 224 | ||
AK098633.1 | - | 3 | 194 | |||
AL136755.2 | - | 3 | 233 | |||
AY626344.1 | - | 3 | 233 | |||
BC033014.1 | - | 3 | 229 | 4827705 | ||
BC047406.1 | - | 3 | 224 | 5298160 | ||
BU570274.1 | - | 3 | 264 | |||
CR533505.1 | - | 2 | 118 |