Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP3A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202855.2 | NP_001189784.1 | 13 | 1569 | ||
NM_017460.5 | NP_059488.2 | 13 | 1572 | |||
GenBank mRNA | AF182273.1 | - | 13 | 1474 | ||
AK298451.1 | - | 12 | 1443 | |||
AK298462.1 | - | 11 | 1319 | |||
AK312967.1 | - | 13 | 1565 | |||
AY606313.2 | - | 13 | 1466 | |||
BC069418.1 | - | 13 | 1515 | 7262313 | ||
BC101631.1 | - | 13 | 1561 | 8069137 | ||
D00003.1 | - | 13 | 1528 | |||
DQ924960.1 | - | 13 | 1465 | |||
J04449.1 | - | 13 | 1530 | |||
M13785.1 | - | 13 | 1528 | |||
M14096.1 | - | 13 | 1533 | |||
M18907.1 | - | 13 | 1555 | |||
X12387.1 | - | 13 | 1534 |