Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4orf17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032149.2 | NP_115525.2 | 5 | 843 | ||
XM_011532315.1 | XP_011530617.1 | 5 | 823 | |||
GenBank mRNA | AK093085.1 | - | 5 | 835 | ||
AK223345.1 | - | 5 | 830 | |||
AK314711.1 | - | 5 | 826 | |||
AL136838.1 | - | 5 | 804 | |||
BC074759.2 | - | 4 | 520 | 30915218 | ||
CR533542.1 | - | 4 | 481 |