Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4orf17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032149.2 | NP_115525.2 | 2 | 444 | ||
XM_011532315.1 | XP_011530617.1 | 2 | 424 | |||
GenBank mRNA | AK093085.1 | - | 2 | 436 | ||
AK223345.1 | - | 2 | 431 | |||
AK314711.1 | - | 2 | 427 | |||
AL136838.1 | - | 2 | 405 | |||
BC074759.2 | - | 1 | 121 | 30915218 | ||
CR533542.1 | - | 1 | 82 | |||
DB051883.1 | - | 2 | 444 |