Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168378.1 | NP_001161850.1 | 2 | 379 | ||
NM_001168379.1 | NP_001161851.1 | 2 | 206 | |||
NM_001243256.1 | NP_001230185.1 | 2 | 505 | |||
NM_032153.5 | NP_115529.2 | 2 | 505 | |||
XM_017007314.1 | XP_016862803.1 | 2 | 2988 | |||
GenBank mRNA | AK056262.1 | - | 2 | 316 | ||
AK293981.1 | - | 2 | 206 | |||
AK294831.1 | - | 2 | 379 | |||
BC029507.1 | - | 2 | 284 | |||
BC126267.1 | - | 2 | 141 | 8991983 | ||
BC136339.1 | - | 2 | 141 | 9020326 | ||
DA066220.1 | - | 2 | 234 | |||
DA218617.1 | - | 2 | 223 | |||
DA759114.1 | - | 2 | 487 |