Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168378.1 | NP_001161850.1 | 3 | 907 | ||
NM_001168379.1 | NP_001161851.1 | 3 | 734 | |||
NM_032153.5 | NP_115529.2 | 3 | 1033 | |||
XM_017007314.1 | XP_016862803.1 | 3 | 3516 | |||
GenBank mRNA | AF332509.1 | - | 1 | 1426 | ||
AK056262.1 | - | 3 | 844 | |||
AK056394.1 | - | 1 | 1218 | |||
AK094286.1 | - | 1 | 530 | |||
AK293981.1 | - | 3 | 734 | |||
AK294831.1 | - | 3 | 907 | |||
BC029507.1 | - | 3 | 812 | |||
BC126267.1 | - | 3 | 669 | 8991983 | ||
BC136339.1 | - | 3 | 669 | 9020326 |