Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCGF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011663.1 | NP_001011663.1 | 8, 9 | 977 | ||
NM_032154.3 | NP_115530.2 | 5, 6 | 752 | |||
GenBank mRNA | AB047006.1 | - | 8, 9 | 931 | ||
AK027885.1 | - | 8, 9 | 953 | |||
AK290679.1 | - | 8, 9 | 931 | |||
AK309105.1 | - | 9, 10 | 1119 | |||
BC007602.1 | - | 5, 6 | 703 | 3350074 | ||
BC010235.1 | - | 8, 9 | 983 | 3461513 |