Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF394 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032164.2 | NP_115540.2 | 3 | 966 | ||
XM_017012711.1 | XP_016868200.1 | 2 | 887 | |||
GenBank mRNA | AK022360.1 | - | 3 | 1009 | ||
AK172865.1 | - | 4 | 887 | |||
AK222948.1 | - | 3 | 959 | |||
AY642122.1 | - | 3 | 993 | |||
BC017051.1 | - | 2 | 849 | |||
BC025241.1 | - | 3 | 993 | 4641268 | ||
BC051847.1 | - | 3 | 966 | 6007172 | ||
L14843.1 | - | 1 | 416 |