Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRIQ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079910.1 | NP_001073379.1 | 12 | 3091 | ||
XM_011538817.2 | XP_011537119.1 | 12 | 3034 | |||
XM_011538818.2 | XP_011537120.1 | 12 | 3034 | |||
XM_011538819.2 | XP_011537121.1 | 12 | 3034 | |||
XM_011538823.2 | XP_011537125.1 | 12 | 3034 | |||
XM_011538824.2 | XP_011537126.1 | 12 | 3034 | |||
XM_011538825.2 | XP_011537127.1 | 12 | 3034 | |||
XM_017020019.1 | XP_016875508.1 | 12 | 3034 | |||
GenBank mRNA | AB058704.1 | - | 8 | 2655 | ||
AK307799.1 | - | 12 | 3042 |