Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303096.1 | NP_001290025.1 | 14 | 1490 | ||
NM_032168.2 | NP_115544.1 | 13 | 1456 | |||
GenBank mRNA | AK022581.1 | - | 13 | 1403 | ||
AK022607.1 | - | 13 | 1415 | |||
AK023276.1 | - | 10 | 1095 | |||
AK091546.1 | - | 14 | 1417 | |||
AK290445.1 | - | 13 | 1407 | |||
AL122097.1 | - | 9 | 1023 | |||
BC040567.1 | - | 13 | 1456 | 5267120 | ||
CR749440.1 | - | 13 | 3065 |