Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032172.2 | NP_115548.1 | 13 | 1821 | ||
XM_005249883.4 | XP_005249940.1 | 13 | 1768 | |||
XM_006715790.1 | XP_006715853.1 | 13 | 1768 | |||
XM_006715791.2 | XP_006715854.1 | 16 | 2274 | |||
XM_011515573.1 | XP_011513875.1 | 17 | 2333 | |||
XM_011515574.1 | XP_011513876.1 | 14 | 1857 | |||
XM_011515577.1 | XP_011513879.1 | 12 | 1486 | |||
XM_011515578.1 | XP_011513880.1 | 14 | 1908 | |||
GenBank mRNA | AJ601395.1 | - | 12 | 1698 | ||
AK022759.1 | - | 13 | 1765 | |||
AK301527.1 | - | 11 | 1293 | |||
AY618868.1 | - | 13 | 1765 | |||
BC060846.1 | - | 13 | 1760 | |||
BC132862.1 | - | 12 | 1718 | 40146884 | ||
BC136656.1 | - | 11 | 2651 |