Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032172.2 | NP_115548.1 | 11 | 1309 | ||
XM_005249883.4 | XP_005249940.1 | 11 | 1256 | |||
XM_006715790.1 | XP_006715853.1 | 11 | 1256 | |||
XM_006715791.2 | XP_006715854.1 | 14 | 1762 | |||
XM_011515573.1 | XP_011513875.1 | 15 | 1821 | |||
XM_011515574.1 | XP_011513876.1 | 12 | 1345 | |||
XM_011515577.1 | XP_011513879.1 | 10 | 974 | |||
XM_011515578.1 | XP_011513880.1 | 12 | 1396 | |||
GenBank mRNA | AJ601395.1 | - | 10 | 1186 | ||
AK022759.1 | - | 11 | 1253 | |||
AK301527.1 | - | 9 | 781 | |||
AK309375.1 | - | 10 | 1050 | |||
AY618868.1 | - | 11 | 1253 | |||
BC060846.1 | - | 11 | 1248 | |||
BC132862.1 | - | 10 | 1206 | 40146884 | ||
BC136656.1 | - | 9 | 2139 |