Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A6OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032178.2 | NP_115554.2 | 3, 4 | 725 | ||
GenBank mRNA | AK023353.1 | - | 3, 4 | 705 | ||
AK091674.1 | - | 2, 3 | 827 | |||
AK225712.1 | - | 3, 4 | 706 | |||
AY078070.1 | - | 3, 4 | 712 | |||
BC013778.1 | - | 3, 4 | 691 | 4345035 |