Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM161A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201543.1 | NP_001188472.1 | 3 | 811 | ||
NM_032180.2 | NP_115556.2 | 3 | 811 | |||
NR_037710.1 | 3 | 755 | ||||
XM_017005072.1 | XP_016860561.1 | 3 | 764 | |||
XM_017005073.1 | XP_016860562.1 | 3 | 738 | |||
XM_017005074.1 | XP_016860563.1 | 3 | 711 | |||
GenBank mRNA | AK023367.1 | - | 4 | 970 | ||
AK296255.1 | - | 2 | 345 | |||
BC107162.1 | - | 3 | 583 | |||
BC107163.1 | - | 3 | 583 | |||
BX648834.1 | - | 3 | 721 | |||
BX649029.1 | - | 3 | 722 |