Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM175A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139076.2 | NP_620775.2 | 5 | 380 | ||
XM_005263280.4 | XP_005263337.1 | 6 | 513 | |||
XM_017008694.1 | XP_016864183.1 | 4 | 286 | |||
GenBank mRNA | AK022704.1 | - | 5 | 366 | ||
AK023676.1 | - | 4 | 280 | |||
AY358576.1 | - | 5 | 331 | |||
BC039573.1 | - | 5 | 327 | 6045433 | ||
DA765424.1 | - | 5 | 380 | |||
EF531340.1 | - | 5 | 315 |