Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM175A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139076.2 | NP_620775.2 | 4 | 307 | ||
XM_005263280.4 | XP_005263337.1 | 5 | 440 | |||
GenBank mRNA | AK022704.1 | - | 4 | 293 | ||
AY358576.1 | - | 4 | 258 | |||
BC039573.1 | - | 4 | 254 | 6045433 | ||
DA765424.1 | - | 4 | 307 | |||
EF531340.1 | - | 4 | 242 |