Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242464.1 | NP_001229393.1 | 5 | 467 | ||
NM_032192.3 | NP_115568.2 | 5 | 892 | |||
NM_181505.3 | NP_852606.1 | 5 | 551 | |||
XM_017025216.1 | XP_016880705.1 | 6 | 1243 | |||
XM_017025217.1 | XP_016880706.1 | 5 | 363 | |||
GenBank mRNA | AF233349.1 | - | 1 | 171 | ||
AF435972.1 | - | 5 | 851 | |||
AF435973.1 | - | 5 | 465 | |||
AF435974.1 | - | 5 | 550 | |||
AF464196.1 | - | 5 | 890 | |||
AK024593.1 | - | 5 | 550 | |||
AK123112.1 | - | 5 | 865 | |||
AK123950.1 | - | 1 | 1927 | |||
AK124417.1 | - | 1 | 1086 | |||
AK127542.1 | - | 6 | 756 | |||
AK129537.1 | - | 4 | 709 | |||
AK225058.1 | - | 5 | 550 | |||
AY070271.1 | - | 5 | 551 | |||
BC001519.2 | - | 5 | 528 | 2987943 | ||
BT007076.1 | - | 4 | 315 | |||
BX648641.1 | - | 2 | 340 |