Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032194.2 | NP_115570.1 | 3, 4 | 276 | ||
GenBank mRNA | AK024740.1 | - | 3, 4 | 253 | ||
AK292165.1 | - | 3, 4 | 255 | |||
AK301662.1 | - | 3, 4 | 245 | |||
BC035314.1 | - | 3, 4 | 205 | 4745524 | ||
BG529605.1 | - | 3, 4 | 277 |