Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MELK SIRNA Assays ›
Gene Symbol:
MELK
Gene Name:
maternal embryonic leucine zipper kinase
Gene Aliases:
HPK38
Chromosome Location:
Chr.9: 36572862 - 36677683 on Build GRCh38
UniGene ID:
Hs.184339
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s386
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MELK
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256685.1
NP_001243614.1
10
939
NM_001256687.1
NP_001243616.1
9
795
NM_001256688.1
NP_001243617.1
8
726
NM_001256689.1
NP_001243618.1
9
853
NM_001256690.1
NP_001243619.1
8
736
NM_001256691.1
NP_001243620.1
8
709
NM_001256692.1
NP_001243621.1
7
613
NM_001256693.1
NP_001243622.1
8
753
NM_014791.3
NP_055606.1
10
939
NR_046337.1
10
976
XM_011518076.2
XP_011516378.1
10
1118
XM_011518077.1
XP_011516379.1
11
1014
XM_011518078.2
XP_011516380.1
11
1201
XM_011518079.1
XP_011516381.1
10
891
XM_011518080.1
XP_011516382.1
10
906
XM_011518081.2
XP_011516383.1
10
924
XM_011518082.2
XP_011516384.1
9
1032
XM_011518083.2
XP_011516385.1
10
1115
XM_011518084.2
XP_011516386.1
9
727
XM_011518085.1
XP_011516387.1
8
723
XM_011518086.2
XP_011516388.1
10
906
GenBank mRNA
AB183427.1
-
9
804
AB183428.1
-
8
689
AK222811.1
-
10
938
AK293284.1
-
8
691
AK293423.1
-
8
674
AK293447.1
-
8
693
AK296820.1
-
10
975
AK299164.1
-
10
891
AK300761.1
-
9
760
AK300821.1
-
9
807
AK301131.1
-
8
685
AK302374.1
-
7
580
AK309470.1
-
9
812
BC014039.2
-
10
893
4547136
D79997.1
-
10
925
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00207681_m1
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
apoptotic process
cell proliferation
intrinsic apoptotic signaling pathway in response to oxidative stress
peptidyl-tyrosine phosphorylation
hemopoiesis
intracellular signal transduction
positive regulation of apoptotic process
protein autophosphorylation
neural precursor cell proliferation
protein serine/threonine kinase activity
non-membrane spanning protein tyrosine kinase activity
calcium ion binding
protein binding
ATP binding
lipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-ndlhf:80/100.66.76.213:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline