Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP7A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000780.3 | NP_000771.2 | 3 | 515 | ||
GenBank mRNA | BC101777.1 | - | 3 | 515 | 8069283 | |
BC112184.1 | - | 3 | 515 | 8327652 | ||
M93133.1 | - | 3 | 511 | |||
X56088.1 | - | 3 | 491 |