Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM188B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032222.2 | NP_115598.2 | 4 | 522 | ||
GenBank mRNA | AF538962.1 | - | 4 | 515 | ||
AK026027.1 | - | 4 | 522 | |||
AK128558.1 | - | 2 | 2706 | |||
BC098569.1 | - | 4 | 469 | 30924492 | ||
DA868176.1 | - | 4 | 522 |
Gene Symbol: INMT-FAM188B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037598.1 | 6 | 974 | |||
GenBank mRNA | AK308762.1 | - | 6 | 972 |