Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289119.1 | NP_001276048.1 | 9 | 1571 | ||
NM_001289120.1 | NP_001276049.1 | 9 | 1594 | |||
NM_001289121.1 | NP_001276050.1 | 9 | 1712 | |||
NM_032226.2 | NP_115602.2 | 9 | 1567 | |||
NR_110317.1 | 8 | 936 | ||||
XM_005251608.4 | XP_005251665.1 | 9 | 2476 | |||
XM_005251612.2 | XP_005251669.1 | 9 | 1534 | |||
XM_011518050.1 | XP_011516352.1 | 8 | 1466 | |||
XM_011518051.2 | XP_011516353.1 | 8 | 6499 | |||
XM_017015197.1 | XP_016870686.1 | 8 | 1593 | |||
XM_017015198.1 | XP_016870687.1 | 8 | 2079 | |||
XM_017015199.1 | XP_016870688.1 | 8 | 1492 | |||
XM_017015200.1 | XP_016870689.1 | 8 | 1765 | |||
XM_017015201.1 | XP_016870690.1 | 8 | 911 | |||
GenBank mRNA | AK074608.1 | - | 8 | 1056 | ||
AK303193.1 | - | 8 | 845 | |||
AK315128.1 | - | 9 | 1584 | |||
BC036940.1 | - | 9 | 1685 | 5399765 |