Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319675.1 | NP_001306604.1 | 3 | 670 | ||
NM_032230.2 | NP_115606.2 | 3 | 578 | |||
XM_005269186.4 | XP_005269243.2 | 3 | 578 | |||
XM_011538827.2 | XP_011537129.1 | 2 | 366 | |||
XM_011538828.2 | XP_011537130.1 | 3 | 578 | |||
XM_011538829.2 | XP_011537131.1 | 3 | 578 | |||
XM_017020020.1 | XP_016875509.1 | 4 | 641 | |||
XM_017020021.1 | XP_016875510.1 | 3 | 1561 | |||
GenBank mRNA | AK026442.1 | - | 3 | 535 | ||
AK310231.1 | - | 3 | 546 | |||
BC029120.1 | - | 3 | 530 | 5189049 |