Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM96A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014812.2 | NP_001014812.1 | 1, 2 | 366 | ||
NM_001289108.1 | NP_001276037.1 | 1, 2 | 366 | |||
NM_032231.5 | NP_115607.1 | 1, 2 | 366 | |||
GenBank mRNA | CN421575.1 | - | 1, 2 | 294 |