Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM96A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014812.2 | NP_001014812.1 | 2 | 478 | ||
NM_001289108.1 | NP_001276037.1 | 2 | 478 | |||
NM_032231.5 | NP_115607.1 | 2 | 478 | |||
NR_110310.1 | 3 | 596 | ||||
GenBank mRNA | AK304202.1 | - | 2 | 265 | ||
AK312171.1 | - | 2 | 420 | |||
BC008865.2 | - | 2 | 380 | 4128736 | ||
BM706342.1 | - | 2 | 478 | |||
BQ269528.1 | - | 2 | 466 | |||
CN421575.1 | - | 2 | 406 | |||
DC415981.1 | - | 3 | 521 |