Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP11A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000781.2 | NP_000772.2 | 9 | 1650 | ||
NM_001099773.1 | NP_001093243.1 | 9 | 1724 | |||
GenBank mRNA | AK056794.1 | - | 9 | 1719 | ||
AK225715.1 | - | 8 | 1238 | |||
AK292300.1 | - | 9 | 1556 | |||
AK292301.1 | - | 8 | 1514 | |||
AK293211.1 | - | 10 | 1516 | |||
AK300178.1 | - | 9 | 1466 | |||
BC032329.1 | - | 9 | 1539 | 5165263 | ||
KU178011.1 | - | 9 | 1495 | |||
M14565.1 | - | 9 | 1539 | |||
M28253.1 | - | 5 | 649 |