Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP11A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000781.2 | NP_000772.2 | 2 | 556 | ||
NM_001099773.1 | NP_001093243.1 | 2 | 630 | |||
GenBank mRNA | AK225715.1 | - | 2 | 462 | ||
AK292300.1 | - | 2 | 462 | |||
AK293204.1 | - | 2 | 462 | |||
AK293211.1 | - | 2 | 462 | |||
AK300178.1 | - | 2 | 462 | |||
AK310362.1 | - | 2 | 462 | |||
BC032329.1 | - | 2 | 445 | 5165263 | ||
BX398579.2 | - | 2 | 556 | |||
KU178011.1 | - | 2 | 401 | |||
M14565.1 | - | 2 | 445 |