Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEX3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032246.4 | NP_115622.2 | 1, 2 | 698 | ||
GenBank mRNA | AK131424.1 | - | 1, 2 | 575 | ||
AY950678.1 | - | 1, 2 | 260 | |||
BC036211.1 | - | 1, 2 | 696 | 5266259 | ||
BC111545.1 | - | 1, 2 | 674 | 5742971 |