Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP11A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000781.2 | NP_000772.2 | 4 | 968 | ||
NM_001099773.1 | NP_001093243.1 | 4 | 1042 | |||
GenBank mRNA | AK056794.1 | - | 4 | 1037 | ||
AK292300.1 | - | 4 | 874 | |||
AK292301.1 | - | 3 | 832 | |||
AK293204.1 | - | 3 | 674 | |||
AK293211.1 | - | 5 | 834 | |||
AK300178.1 | - | 4 | 784 | |||
AY603498.1 | - | 1 | 17 | |||
BC032329.1 | - | 4 | 857 | 5165263 | ||
KU178011.1 | - | 4 | 813 | |||
M14565.1 | - | 4 | 857 |